Assay Details
| Gene Symbol: CNST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152609.2 | NP_689822.2 | 11 | 3878 | 93 | |
| XM_005273081.3 | XP_005273138.2 | 11 | 3663 | 93 | ||
| XM_005273083.4 | XP_005273140.1 | 10 | 3581 | 93 | ||
| XM_011544110.2 | XP_011542412.1 | 11 | 3729 | 93 | ||
| XM_011544111.1 | XP_011542413.1 | 11 | 3900 | 93 | ||
| XM_011544112.1 | XP_011542414.1 | 10 | 3447 | 93 | ||
| XM_011544113.1 | XP_011542415.1 | 10 | 3572 | 93 | ||
| XM_011544114.2 | XP_011542416.1 | 10 | 3287 | 93 | ||
| XM_017000487.1 | XP_016855976.1 | 12 | 3918 | 93 | ||
| XM_017000488.1 | XP_016855977.1 | 11 | 3661 | 93 | ||
| XM_017000489.1 | XP_016855978.1 | 11 | 3728 | 93 | ||
| GenBank mRNA | AK074142.1 | - | 11 | 3644 | 93 | |
| AL834246.2 | - | 11 | 3712 | 93 | ||
| BX647629.1 | - | 11 | 3663 | 93 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |

CNST
Genomic Map