Assay Details
| Gene Symbol: SLAMF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017002756.1 | XP_016858245.1 | 1 | 832 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017002756.1 transcripts.
Set Membership: |
|

SLAMF9
Genomic Map