| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006734.hg.1 |
ENST00000526456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006734.hg.1 |
ENST00000526456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006734.hg.1 |
ENST00000526456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006734.hg.1 |
ENST00000526456.1 |
Ensembl |