| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016236.hg.1 |
ENST00000446979.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016236.hg.1 |
ENST00000446979.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016236.hg.1 |
ENST00000446979.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016236.hg.1 |
NR_037809.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016236.hg.1 |
NR_037809.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016236.hg.1 |
NR_037809.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016236.hg.1 |
NR_037809.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016236.hg.1 |
ENST00000446979.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016236.hg.1 |
ENST00000446979.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016236.hg.1 |
ENST00000446979.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016236.hg.1 |
NR_037809.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016236.hg.1 |
NR_037809.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016236.hg.1 |
NR_037809.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016236.hg.1 |
NR_037809.1 |
Refseq |