Assay Details
| Gene Symbol: BCR-PDGFRA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_BCR-PDGFRA_B12ins12P12.0 | - | 116 | 79 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BCR-PDGFRA fusion| Gene Symbol: BCR-PDGFRA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_BCR-PDGFRA_B12ins12P12.0 | - | 116 | 79 | ||