Assay Details
| Gene Symbol: ESR1-CCDC170 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ESR1-CCDC170_E2C8.0 | - | 115 | 146 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ESR1-CCDC170 fusion| Gene Symbol: ESR1-CCDC170 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ESR1-CCDC170_E2C8.0 | - | 115 | 146 | ||
Set Membership: |
|