Assay Details
| Gene Symbol: FGFR2-SHTN1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-SHTN1_F17S7.0 | - | 114 | 122 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGFR2-SHTN1 fusion| Gene Symbol: FGFR2-SHTN1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-SHTN1_F17S7.0 | - | 114 | 122 | ||
Set Membership: |
|