Assay Details
| Gene Symbol: FN1-FGFR1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FN1-FGFR1_F23F3.0 | - | 117 | 91 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FN1-FGFR1 fusion| Gene Symbol: FN1-FGFR1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FN1-FGFR1_F23F3.0 | - | 117 | 91 | ||