Assay Details
| Gene Symbol: NF1-ACACA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NF1-ACACA_N1A42.0 | - | 21 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NF1-ACACA fusion| Gene Symbol: NF1-ACACA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NF1-ACACA_N1A42.0 | - | 21 | 100 | ||