Assay Details
| Gene Symbol: NF1-ATAD5 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NF1-ATAD5_N5A11.0 | - | 115 | 90 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NF1-ATAD5 fusion| Gene Symbol: NF1-ATAD5 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NF1-ATAD5_N5A11.0 | - | 115 | 90 | ||