Assay Details
| Gene Symbol: PCNX-RAD51B fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PCNX-RAD51B_P1R8.0 | - | 123 | 102 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCNX-RAD51B fusion| Gene Symbol: PCNX-RAD51B fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PCNX-RAD51B_P1R8.0 | - | 123 | 102 | ||
Set Membership: |
|