Assay Details
| Gene Symbol: RNF213-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_RNF213-ALK_R20A20.0 | - | 117 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF213-ALK fusion| Gene Symbol: RNF213-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_RNF213-ALK_R20A20.0 | - | 117 | 82 | ||