Assay Details
| Gene Symbol: SEC61G-EGFR fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SEC61G-EGFR_S2E9.0 | - | 111 | 126 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEC61G-EGFR fusion| Gene Symbol: SEC61G-EGFR fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SEC61G-EGFR_S2E9.0 | - | 111 | 126 | ||
Set Membership: |
|