Assay Details
| Gene Symbol: BCAM-AKT2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_BCAM-AKT2_B13A5.0 | - | 67 | 96 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BCAM-AKT2 fusion| Gene Symbol: BCAM-AKT2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_BCAM-AKT2_B13A5.0 | - | 67 | 96 | ||