Assay Details
| Gene Symbol: FGFR2-CCDC6 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-CCDC6_F17C2.0 | - | 52 | 129 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGFR2-CCDC6 fusion| Gene Symbol: FGFR2-CCDC6 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-CCDC6_F17C2.0 | - | 52 | 129 | ||
Set Membership: |
|