Assay Details
| Gene Symbol: FGFR2-USP10 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-USP10_F17del11U5.0 | - | 73 | 86 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGFR2-USP10 fusion| Gene Symbol: FGFR2-USP10 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-USP10_F17del11U5.0 | - | 73 | 86 | ||