Assay Details
| Gene Symbol: GATM-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_GATM-BRAF_G2B11.0 | - | 73 | 72 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GATM-BRAF fusion| Gene Symbol: GATM-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_GATM-BRAF_G2B11.0 | - | 73 | 72 | ||