Assay Details
| Gene Symbol: GOLGA4-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_GOLGA4-PDGFRB_G10P11.0 | - | 44 | 85 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GOLGA4-PDGFRB fusion| Gene Symbol: GOLGA4-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_GOLGA4-PDGFRB_G10P11.0 | - | 44 | 85 | ||