Assay Details
| Gene Symbol: MIR143HG-NOTCH1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MIR143HG-NOTCH1_M1N27.0 | - | 70 | 106 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR143HG-NOTCH1 fusion| Gene Symbol: MIR143HG-NOTCH1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MIR143HG-NOTCH1_M1N27.0 | - | 70 | 106 | ||
Set Membership: |
|