Assay Details
| Gene Symbol: MKRN1-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MKRN1-BRAF_M4B9.0 | - | 64 | 79 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MKRN1-BRAF fusion| Gene Symbol: MKRN1-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MKRN1-BRAF_M4B9.0 | - | 64 | 79 | ||