Assay Details
| Gene Symbol: MYBL1-NFIB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYBL1-NFIB_M14N11.0 | - | 65 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYBL1-NFIB fusion| Gene Symbol: MYBL1-NFIB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYBL1-NFIB_M14N11.0 | - | 65 | 97 | ||