Assay Details
| Gene Symbol: MYBL1-NFIB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYBL1-NFIB_M15N11.0 | - | 67 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYBL1-NFIB fusion| Gene Symbol: MYBL1-NFIB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYBL1-NFIB_M15N11.0 | - | 67 | 101 | ||
Set Membership: |
|