Assay Details
| Gene Symbol: MYB-QKI fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYB-QKI_M9Q5.0 | - | 45 | 96 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYB-QKI fusion| Gene Symbol: MYB-QKI fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYB-QKI_M9Q5.0 | - | 45 | 96 | ||