Assay Details
| Gene Symbol: NF1-PSMD11 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NF1-PSMD11_N5P2.0 | - | 65 | 125 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NF1-PSMD11 fusion| Gene Symbol: NF1-PSMD11 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NF1-PSMD11_N5P2.0 | - | 65 | 125 | ||
Set Membership: |
|