Assay Details
| Gene Symbol: NOTCH1-SNHG7 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NOTCH1-SNHG7_N2S4.0 | - | 79 | 89 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NOTCH1-SNHG7 fusion| Gene Symbol: NOTCH1-SNHG7 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NOTCH1-SNHG7_N2S4.0 | - | 79 | 89 | ||