Assay Details
| Gene Symbol: PCM1-JAK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PCM1-JAK2_P23J12_COSF1001.0 | - | 54 | 114 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCM1-JAK2 fusion| Gene Symbol: PCM1-JAK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PCM1-JAK2_P23J12_COSF1001.0 | - | 54 | 114 | ||
Set Membership: |
|