Assay Details
| Gene Symbol: PPL-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PPL-NTRK1_P22N11.0 | - | 50 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPL-NTRK1 fusion| Gene Symbol: PPL-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PPL-NTRK1_P22N11.0 | - | 50 | 97 | ||