Assay Details
| Gene Symbol: PTEN-SHROOM4 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PTEN-SHROOM4_P2S3.0 | - | 75 | 95 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTEN-SHROOM4 fusion| Gene Symbol: PTEN-SHROOM4 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PTEN-SHROOM4_P2S3.0 | - | 75 | 95 | ||