Assay Details
| Gene Symbol: SQSTM1-FGFR1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SQSTM1-FGFR1_S6F10.0 | - | 48 | 95 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SQSTM1-FGFR1 fusion| Gene Symbol: SQSTM1-FGFR1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SQSTM1-FGFR1_S6F10.0 | - | 48 | 95 | ||