Assay Details
| Gene Symbol: SSBP2-JAK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SSBP2-JAK2_S4J11_COSF1047.0 | - | 62 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SSBP2-JAK2 fusion| Gene Symbol: SSBP2-JAK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SSBP2-JAK2_S4J11_COSF1047.0 | - | 62 | 100 | ||