Assay Details
| Gene Symbol: TP53BP1-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TP53BP1-PDGFRB_T23P11.0 | - | 67 | 105 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TP53BP1-PDGFRB fusion| Gene Symbol: TP53BP1-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TP53BP1-PDGFRB_T23P11.0 | - | 67 | 105 | ||
Set Membership: |
|