Assay Details
| Gene Symbol: CHTOP-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CHTOP-NTRK1_C5N10.0 | - | 53 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHTOP-NTRK1 fusion| Gene Symbol: CHTOP-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CHTOP-NTRK1_C5N10.0 | - | 53 | 94 | ||