Assay Details
| Gene Symbol: DIP2C-PDGFRA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_DIP2C-PDGFRA_D1P11.0 | - | 50 | 84 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DIP2C-PDGFRA fusion| Gene Symbol: DIP2C-PDGFRA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_DIP2C-PDGFRA_D1P11.0 | - | 50 | 84 | ||