Assay Details
| Gene Symbol: EGFR-SEPT14 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_EGFR-SEPT14_E24S10.0 | - | 58 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EGFR-SEPT14 fusion| Gene Symbol: EGFR-SEPT14 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_EGFR-SEPT14_E24S10.0 | - | 58 | 97 | ||