Assay Details
| Gene Symbol: EPS15-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_EPS15-BRAF_E22B10.0 | - | 82 | 98 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPS15-BRAF fusion| Gene Symbol: EPS15-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_EPS15-BRAF_E22B10.0 | - | 82 | 98 | ||