Assay Details
| Gene Symbol: ESR1-CCDC170 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ESR1-CCDC170_E2C6.0 | - | 57 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ESR1-CCDC170 fusion| Gene Symbol: ESR1-CCDC170 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ESR1-CCDC170_E2C6.0 | - | 57 | 100 | ||