Assay Details
| Gene Symbol: ESR1-DAB2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ESR1-DAB2_E6D3.0 | - | 95 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ESR1-DAB2 fusion| Gene Symbol: ESR1-DAB2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ESR1-DAB2_E6D3.0 | - | 95 | 94 | ||