Assay Details
| Gene Symbol: ETV6-FGFR3 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ETV6-FGFR3_E5F9.0 | - | 99 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ETV6-FGFR3 fusion| Gene Symbol: ETV6-FGFR3 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ETV6-FGFR3_E5F9.0 | - | 99 | 94 | ||