Assay Details
| Gene Symbol: ETV6-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ETV6-PDGFRB_E4P9.0 | - | 52 | 98 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ETV6-PDGFRB fusion| Gene Symbol: ETV6-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ETV6-PDGFRB_E4P9.0 | - | 52 | 98 | ||