Assay Details
| Gene Symbol: EZR-ERBB4 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_EZR-ERBB4_E12E18.0 | - | 101 | 117 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EZR-ERBB4 fusion| Gene Symbol: EZR-ERBB4 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_EZR-ERBB4_E12E18.0 | - | 101 | 117 | ||
Set Membership: |
|