Assay Details
| Gene Symbol: FGFR1-ADAM32 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR1-ADAM32_F17A14.0 | - | 53 | 81 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGFR1-ADAM32 fusion| Gene Symbol: FGFR1-ADAM32 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR1-ADAM32_F17A14.0 | - | 53 | 81 | ||