Assay Details
| Gene Symbol: FGFR2-FAM76A fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-FAM76A_F17F2.0 | - | 89 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGFR2-FAM76A fusion| Gene Symbol: FGFR2-FAM76A fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-FAM76A_F17F2.0 | - | 89 | 101 | ||
Set Membership: |
|