Assay Details
| Gene Symbol: GLIS3-TERT fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_GLIS3-TERT_G3T3.0 | - | 76 | 110 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLIS3-TERT fusion| Gene Symbol: GLIS3-TERT fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_GLIS3-TERT_G3T3.0 | - | 76 | 110 | ||
Set Membership: |
|