Assay Details
| Gene Symbol: GOLGB1-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_GOLGB1-PDGFRB_G10P12.0 | - | 48 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GOLGB1-PDGFRB fusion| Gene Symbol: GOLGB1-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_GOLGB1-PDGFRB_G10P12.0 | - | 48 | 88 | ||