Assay Details
| Gene Symbol: KTN1-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_KTN1-ALK_K43A19.0 | - | 57 | 96 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KTN1-ALK fusion| Gene Symbol: KTN1-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_KTN1-ALK_K43A19.0 | - | 57 | 96 | ||