Assay Details
| Gene Symbol: PTEN-BTAF1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PTEN-BTAF1_P2B2.0 | - | 67 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTEN-BTAF1 fusion| Gene Symbol: PTEN-BTAF1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PTEN-BTAF1_P2B2.0 | - | 67 | 100 | ||