Assay Details
| Gene Symbol: SNX19-FGFR2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SNX19-FGFR2_S7F7.0 | - | 92 | 87 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNX19-FGFR2 fusion| Gene Symbol: SNX19-FGFR2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SNX19-FGFR2_S7F7.0 | - | 92 | 87 | ||