Assay Details
| Gene Symbol: TSEN2-PPARG fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TSEN2-PPARG_T6P6.0 | - | 48 | 116 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSEN2-PPARG fusion| Gene Symbol: TSEN2-PPARG fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TSEN2-PPARG_T6P6.0 | - | 48 | 116 | ||
Set Membership: |
|