Assay Details
| Gene Symbol: FGFR1OP-FGFR1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR1OP-FGFR1_F6F10.0 | - | 130 | 105 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGFR1OP-FGFR1 fusion| Gene Symbol: FGFR1OP-FGFR1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR1OP-FGFR1_F6F10.0 | - | 130 | 105 | ||
Set Membership: |
|