Assay Details
| Gene Symbol: NSD1-NOTCH4 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NSD1-NOTCH4_N14N18.0 | - | 90 | 90 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NSD1-NOTCH4 fusion| Gene Symbol: NSD1-NOTCH4 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NSD1-NOTCH4_N14N18.0 | - | 90 | 90 | ||